missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoic acid, 94%

Product Code. 15488999
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15488999

Brand: Thermo Scientific Alfa Aesar L14191.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-43-1 | C7H3Cl3O2 | 225.449 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 50-43-1
Melting Point 162°C to 164°C
Molecular Formula C7H3Cl3O2
MDL Number MFCD00060699
Quantity 5 g
Beilstein 1874127
Synonym benzoic acid, 2,4,6-trichloro, unii-0bwg1hx6ck, 2,4,6-trichlorobenzoicacid, 2,4,6-trichloro-benzoic acid, 0bwg1hx6ck, 2,4,6-trichlorbenzoes, pubchem18496, acmc-209kme, 2,4,6-trichlorobbenzoicacid, ksc270e7f
InChI Key RAFFVQBMVYYTQS-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Cl)C(=O)O)Cl)Cl
IUPAC Name 2,4,6-trichlorobenzoic acid
Molecular Weight (g/mol) 225.449
PubChem CID 5764
Formula Weight 225.46
Percent Purity 94%
Chemical Name or Material 2,4,6-Trichlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Trichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4,6-Trichlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.