missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylmethyl)triphenylphosphonium chloride, 97%, Thermo Scientific™

Product Code. 11433368
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11433368

Brand: Thermo Scientific Alfa Aesar A17775.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Very Soluble in water (100 g/L) (25°C).

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-97-7
Molecular Formula C21H20ClO2P
Molecular Weight (g/mol) 370.81
MDL Number MFCD00050199
InChI Key CXCXTEMZMJZMJX-UHFFFAOYSA-M
Synonym carbomethoxymethyl triphenylphosphonium chloride, 2-methoxy-2-oxoethyl triphenylphosphonium chloride, 2-methoxy-2-oxoethyl triphenylphosphanium chloride, carbomethoxymethyltriphenylphosphonium chloride, methoxycarbonylmethyl triphenylphosphonium chloride, acmc-20ali4, carbomethoxymethyltriphenylphosphoniumchloride, methyl 2-triphenylphosphino acetate, chloride, 2-methoxy-2-oxoethyl-triphenylphosphanium chloride, 2-methoxy-2-oxoethyl-triphenylphosphonium chloride
PubChem CID 2733549
SMILES [Cl-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼162°C (decomposition)
Quantity 50 g
Beilstein 4172737
Solubility Information Very Soluble in water (100g/L) (25°C).
IUPAC Name (2-methoxy-2-oxoethyl)triphenylphosphanium chloride
Formula Weight 370.82
Percent Purity 97%
Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.