missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11425596
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11425596

Brand: Thermo Scientific Alfa Aesar B20731.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32890-87-2
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00042496
InChI Key JTOIZLCQNWWDCN-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzoic acid, 2,4-bis-trifluoromethylbenzoic acid, 2,4-di trifluoromethyl benzoic acid, benzoic acid, 2,4-bis trifluoromethyl, 2,4-bis-trifluoromethyl-benzoic acid, maybridge4_003283, acmc-20aoeb, pubchem8491, rarechem al bo 0512
PubChem CID 141752
IUPAC Name 2,4-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)O

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 2057460
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.