Learn More
Alfa Aesar™ 2-(4-Bromophenyl)benzimidazole, 95%
Brand: Alfa Aesar™ H64811.06
Additional Details : CAS : 2622-74-4 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
2622-74-4 | |
273.133 | |
YRWMGOSKROWAIT-UHFFFAOYSA-N | |
286182 | |
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Br |
C13H9BrN2 | |
MFCD00224360 | |
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885 | |
2-(4-bromophenyl)-1H-benzimidazole |
Specifications
2-(4-Bromophenyl)benzimidazole | |
2622-74-4 | |
MFCD00224360 | |
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885 | |
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Br | |
273.133 | |
273.13 | |
95% |
5g | |
C13H9BrN2 | |
162670 | |
YRWMGOSKROWAIT-UHFFFAOYSA-N | |
2-(4-bromophenyl)-1H-benzimidazole | |
286182 | |
Solid |