missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid, 97%

Brand:  Alfa Aesar™ H54240.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410946

  • 202.00€ / 1g


Estimated Shipment
17-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

175401-95-3
263.256
HRUUSKALOILBKF-UHFFFAOYSA-N
23509758
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
C15H9N3O2
MFCD20265213
2-4-cyanophenyl benzimidazole-6-carboxylic acid, 2-4-cyanophenyl benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-3h-1,3-benzodiazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-6-carboxylic acid, 1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl, 2-4-cyanophenyl-1h-benzoimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzo d imidazole-6-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-1,3-benzodiazole-5-carboxylic acid
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid

Specifications

2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid
175401-95-3
MFCD20265213
HRUUSKALOILBKF-UHFFFAOYSA-N
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
23509758
97%
1g
C15H9N3O2
2-4-cyanophenyl benzimidazole-6-carboxylic acid, 2-4-cyanophenyl benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-3h-1,3-benzodiazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-6-carboxylic acid, 1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl, 2-4-cyanophenyl-1h-benzoimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzo d imidazole-6-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-1,3-benzodiazole-5-carboxylic acid
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
263.256
263.26
Videos
SDS Documents