missing translation for 'onlineSavingsMsg'
Learn More

2-([4-(Ethoxycarbonyl)phenyl]dimethylsilyl)benzyl alcohol, 95%, Thermo Scientific™

Product Code. 15419435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419435 1 g 1g
15429435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419435 Supplier Thermo Scientific Alfa Aesar Supplier No. H51932.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1244855-68-2
Molecular Formula C18H22O3Si
Molecular Weight (g/mol) 314.46
MDL Number MFCD16251591
InChI Key VPQQJIOOSRZZDJ-UHFFFAOYSA-N
Synonym ethyl 4-2-hydroxymethyl phenyl dimethylsilyl benzoate, 2-4-ethoxycarbonyl phenyl dimethylsilyl benzyl alcohol, amtsi153, 4-2-hydroxymethyl-phenyl-dimethyl-silanyl-benzoic acid ethyl ester
PubChem CID 71306647
IUPAC Name ethyl 4-[[2-(hydroxymethyl)phenyl]-dimethylsilyl]benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)[Si](C)(C)C1=CC=CC=C1CO

Specifications

Quantity 1 g
Formula Weight 314.46
Percent Purity 95%
Chemical Name or Material 2-([4-(Ethoxycarbonyl)phenyl]dimethylsilyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.