missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 15411386
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
57677-80-2
C11H9NO3S
235.257
MFCD03791177
OIBLFOXWCGIRIQ-UHFFFAOYSA-N
2-4-methoxyphenyl thiazole-4-carboxylic acid, 2-4-methoxyphenyl-1,3-thiazole-4-carboxylic acid, 2-4-methoxy-phenyl-thiazole-4-carboxylic acid, 2-4-methoxyphenyl-4-thiazolecarboxylic acid, acmc-209lzq, 2-4-methoxyphenyl thiazole-4-carboxylicacid, 4-thiazolecarboxylicacid, 2-4-methoxyphenyl, 4-thiazolecarboxylic acid, 2-4-methoxyphenyl
2763565
2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylic acid
COC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
This item is not returnable. View return policy
57677-80-2
C11H9NO3S
235.257
MFCD03791177
OIBLFOXWCGIRIQ-UHFFFAOYSA-N
2-4-methoxyphenyl thiazole-4-carboxylic acid, 2-4-methoxyphenyl-1,3-thiazole-4-carboxylic acid, 2-4-methoxy-phenyl-thiazole-4-carboxylic acid, 2-4-methoxyphenyl-4-thiazolecarboxylic acid, acmc-209lzq, 2-4-methoxyphenyl thiazole-4-carboxylicacid, 4-thiazolecarboxylicacid, 2-4-methoxyphenyl, 4-thiazolecarboxylic acid, 2-4-methoxyphenyl
2763565
2-(4-methoxyphenyl)-1,3-thiazole-4-carboxylic acid
COC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 235.26
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxyphenyl)thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'