missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 2'5' ADP Sepharose 4B binds and immobilizes enzymes requiring NADP cofactor
Specifications
Specifications
| Binding Capacity | >0.4mg G6PDH/mL |
| Concentration | Approx. 2 μmol/mL 2'5' ADP/Drained Medium |
| Ligand Concentration | 2μM 2′5′ ADP/mL Drained Medium |
| Matrix | 4% Agarose |
| Particle Size | 90 μm |
| pH | 4 to 10 |
| Quantity | 5 g |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Used for purification of Enzymes requiring NADP as a cofactor |
| Material | 4% Agarose |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?