missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-n-octylthiophene, 96%, Thermo Scientific™

Catalog Number 15411228
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
149703-84-4
C12H18Br2S
354.14
MFCD00672013
PJGBSUPPENVFAD-UHFFFAOYSA-N
2,5-dibromo-3-n-octylthiophene, 2,5-dibromo-3-octyl-thiophene, acmc-1c21j, thiophene, 2,5-dibromo-3-octyl, 2,5-bis bromanyl-3-octyl-thiophene, 2,5-dibromo-3-octylthiophene
4385790
2,5-dibromo-3-octylthiophene
CCCCCCCCC1=C(Br)SC(Br)=C1
This item is not returnable. View return policy
149703-84-4
C12H18Br2S
354.14
MFCD00672013
PJGBSUPPENVFAD-UHFFFAOYSA-N
2,5-dibromo-3-n-octylthiophene, 2,5-dibromo-3-octyl-thiophene, acmc-1c21j, thiophene, 2,5-dibromo-3-octyl, 2,5-bis bromanyl-3-octyl-thiophene, 2,5-dibromo-3-octylthiophene
4385790
2,5-dibromo-3-octylthiophene
CCCCCCCCC1=C(Br)SC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a conducting polymer precursor.

Solubility
Slightly soluble in water.

Notes
Store at 2-8°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.44
Boiling Point 316°C
Flash Point 110°C (230°F)
Refractive Index 1.548
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 354.14
Percent Purity 96%
Chemical Name or Material 2,5-Dibromo-3-n-octylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'