missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-3757623
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
135145-90-3
C6H5BCl2O2
190.82
MFCD01863182
NNTFPBXQPOQRBT-UHFFFAOYSA-N
2,5-dichlorobenzeneboronic acid, 2,5-dichlorophenyl boronic acid, 2,5-dichlorophenylboronicacid, 2,5-dichlorophenyl boranediol, boronic acid, 2,5-dichlorophenyl, pubchem1811, acmc-209bxy, ksc174k3h, 2,5-dichloro-phenylboronic acid
2773371
(2,5-dichlorophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)Cl)(O)O
This item is not returnable. View return policy
135145-90-3
C6H5BCl2O2
190.82
MFCD01863182
NNTFPBXQPOQRBT-UHFFFAOYSA-N
2,5-dichlorobenzeneboronic acid, 2,5-dichlorophenyl boronic acid, 2,5-dichlorophenylboronicacid, 2,5-dichlorophenyl boranediol, boronic acid, 2,5-dichlorophenyl, pubchem1811, acmc-209bxy, ksc174k3h, 2,5-dichloro-phenylboronic acid
2773371
(2,5-dichlorophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)Cl)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 190.82
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 5-Dichlorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichlorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'