missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pseudomonas Agar Base (Dehydrated)

Product Code. 12731560
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 12731560

Brand: Thermo Scientific™ CM0559R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively isolate Pseudomonasspecies from a range of samples with this medium.

Selectively isolate Pseudomonas species from a range of samples with Thermo Scientific™ Oxoid™ Pseudomonas Agar Base (Dehydrated). The medium is modification of King's A Medium1 and contains magnesium chloride and potassium sulphate for improved pigment production and recovery of Pseudomonas aeruginosa while strongly suppressing Klebsiella, Proteus and Providencia spp.

Pseudomonas Agar Base is designed so that by the addition of the appropriate supplement (SR0102 or SR0103) the medium becomes selective for Pseudomonas aeruginosa or Pseudomonas spp. generally.

  • Selective: Pseudomonas aeruginosa forms fluorescent green colonies while the growth of Klebsiella, Proteus, and Providencia is strongly suppressed
  • Enhanced pigment production aids identification

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Agar
Quantity 2.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.