missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol, 97%, Thermo Scientific™

Catalog Number p-2696325
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
103788-65-4
C12H13NO2
203.241
MFCD00100006
JYWHQBLLIBQGCU-UHFFFAOYSA-N
725585
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol
CC1=C(N=C(O1)C2=CC=CC=C2)CCO
This item is not returnable. View return policy
103788-65-4
C12H13NO2
203.241
MFCD00100006
JYWHQBLLIBQGCU-UHFFFAOYSA-N
725585
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol
CC1=C(N=C(O1)C2=CC=CC=C2)CCO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethan-1-ol, 2-5-methyl-2-phenyloxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyloxazol, 4-oxazoleethanol, 5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl-ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyl-1,3-oxazole, 4-oxazoleethanol,5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl-1-ethanol
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)ethan-1-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'