missing translation for 'onlineSavingsMsg'
Learn More

Reagecon 2.5LT POT.BR/KBRO3 0,167M 2,5L ANALYTICAL VOLUMETRIC SO

Brand:  Reagecon KB20016W


2.5LT POT.BR/KBRO3 0,167M 2,5L ANALYTICAL VOLUMETRIC SO

Product Code. 11506823

  • 103.00€ / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • POTASSIUM BROMIDE / BROMATE 0.1N (0.0167M) Analytical Volumetric Solution

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : potassium bromate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.