missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Phosphate-d34, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
Specifications
Specifications
| Chemical Name or Material | Bis(2-ethylhexyl) Phosphate-d34 |
| CAS | 1773493-20-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 D34 O4 P |
| Quantity | 2.5 mg |
| Synonym | 2-Ethyl-1-hexanol Hydrogen Phosphate-d34, 2-Ethyl-1-hexanol Hydrogen Phosphate A 208-d34, A 208 (phosphate)-d34, Baysolvex D 2EHPA-d34, Bis(2-ethylhexyl) Hydrogen Phosphate-d34, Bis(2-ethylhexyl) Orthophosphate-d34, Bis(2-ethylhexyl) phosphate-d34, D 2EHPA-d34, DEHAP-d34, DP 8-d34, DP 8R-d34, Di(2-ethylhexyl) Phosphate-d34, Di(2-ethylhexyl)orthophosphoric Acid-d34, Di(2-ethylhexyl)phosphoric Acid-d34, Di-2-ethylhexyl Hydrogen Phosphate-d34, Dioctyl Hydrogen Phosphate-d34, Dioctyl Orthophosphate-d34, Dioctyl Phosphate-d34, HDEHP-d34, HDEHPA-d34, Hostarex PA 216-d34, Hydrogen Bis(2-ethylhexyl) Phosphate-d34, JB 58-d34, LB 58-d34, O,O-Bis(2-ethylhexyl)phosphoric Acid-d34, O,O-Di(2-ethylhexyl)phosphoric Acid-d34, Orthophosphoric Acid 2-Ethylhexyl Alcohol Diester-d34, P 204-d34, P 204 (acid)-d34, Phoslex A 208-d34, Phosphoric Acid di(2-ethylhexyl)Ester-d34, TOPS 99-d34 |
| InChI Formula | InChI=1S/C16H35O4P/c1-5-9-11-15(7-3)13-19-21(17,18)20-14-16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3,(H,17,18)/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D,16D |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])OP(=O)([O-])OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | bis[1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexyl] phosphate |
| Molecular Weight (g/mol) | 355.622 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction