Learn More
Tryptamine-d4 Hydrochloride, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Tryptamine-d4 Hydrochloride |
| CAS | 340257-60-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 2H4 H8 N2 . Cl H |
| Quantity | 2.5 mg |
| Synonym | 1H-Indole-3-ethan-α,α,β,β-d4-amine','monohydrochloride (9CI), 2-(1H-Indol-3-yl)ethanamine-1,1,2,2-d4 hydrochloride, 3-(2-Aminoethyl-d4)-1H-indole hydrochloride, Tryptamine-d4 hydrochloride, Tryptamine-α,α,β,β-d4 hydrochloride, Tryptamine-D4 (aminoethyl-D4) hydrochloride |
| InChI Formula | InChI=1S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12H,5-6,11H2;1H/i5D2,6D2; |
| SMILES | Cl.[2H]C([2H])(N)C([2H])([2H])c1c[nH]c2ccccc12 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(1H-indol-3-yl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 200.7 |
| Show More |