missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-3211561
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
38222-83-2
C14H23N
205.35
MFCD00006305
HVHZEKKZMFRULH-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
98898
2,6-ditert-butyl-4-methylpyridine
CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
This item is not returnable. View return policy
38222-83-2
C14H23N
205.35
MFCD00006305
HVHZEKKZMFRULH-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
98898
2,6-ditert-butyl-4-methylpyridine
CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color Colorless to Yellow
Boiling Point 233°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2C5H2NCH3
Refractive Index 1.4760 to 1.478
Quantity 5 g
Beilstein 20, V,6, 190
Fieser 09,141; 10,123; 11,160; 16,110
Merck Index 15, 3042
Formula Weight 205.34
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylpyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Di-tert-butyl-4-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'