missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidophenol, 97%, Thermo Scientific Chemicals

Catalog Number p-3211567
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
614-80-2
C8H9NO2
151.16
MFCD00002181
ADVGKWPZRIDURE-UHFFFAOYSA-N
2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
11972
N-(2-hydroxyphenyl)acetamide
CC(=O)NC1=CC=CC=C1O
This item is not returnable. View return policy
614-80-2
C8H9NO2
151.16
MFCD00002181
ADVGKWPZRIDURE-UHFFFAOYSA-N
2-acetamidophenol, n-2-hydroxyphenyl acetamide, 2'-hydroxyacetanilide, o-hydroxyacetanilide, 2-acetaminophenol, 2-hydroxyacetanilide, o-acetamidophenol, o-acetylamino phenol, 2-acetylamino phenol, o-acetaminophenol
11972
N-(2-hydroxyphenyl)acetamide
CC(=O)NC1=CC=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 207°C to 210°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3CONHC6H4OH
Quantity 25 g
Beilstein 13,37
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol,alkaline solutions,slightly in methanol
Formula Weight 151.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Acetamidophenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetamidophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'