Learn More
Julabo™ 2-Adapters for Recirculating Cooler
Click to view available options
Connection Type:
G 1 1/4 in. F to 1 in. Barb. Fit
G 1 1/4 in. F to 3/4 in. Barb. Fit
G 1 1/4 in. F to NPT 1 in. M
G 1 1/4 in. F to NPT 1/2 in. M
G 1 1/4 in. F to NPT 3/4 in. M
G 1 1/4 in. Female to M16 x 1 Male
G 3/4 in. F to 1/2 in. Barb. Fit
G 3/4 in. F to NPT 1/2 in. M
G 3/4 in. F to NPT 3/4 in. M
G 3/4 in. Female to Barbed Fitting
G 3/4 in. Female to M16 x 1 Male
For Use With (Equipment):
FL11006,FL20006,FL2506,FL4006,FL7006
FL1203, FL1703, FL2503, FL4003
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | FL1203, FL1703, FL2503, FL4003 |
| Type | 2-Adapters |
| Connection Type | G 3/4 in. F to 1/2 in. Barb. Fit |