missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyrazine, 99%, Thermo Scientific Chemicals

Product Code. 11422130
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422130 1 g 1g
15473827 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422130 Supplier Thermo Scientific Alfa Aesar Supplier No. H28744.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24241-18-7
Molecular Formula C4H3Br2N3
Molecular Weight (g/mol) 252.90
MDL Number MFCD00673150
InChI Key DTLBKXRFWUERQN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyrazine, 3,5-dibromo-2-pyrazinamine, 3,5-dibromopyrazin-2-ylamine, 3,5-dibromopyrazine-2-ylamine, amino-3,5-dibromopyrazine, pyrazinamine, 3,5-dibromo, 3,5-dibromopyrazine-2-amine, 2-amino-3,5-dibromo pyrazine, 2-amino-3,5-dibromo-pyrazine, 2-pyrazinamine, 3,5-dibromo
PubChem CID 620004
IUPAC Name 3,5-dibromopyrazin-2-amine
SMILES NC1=NC=C(Br)N=C1Br

Specifications

Melting Point 114°C to 117°C
Quantity 1 g
Formula Weight 252.9
Percent Purity 99%
Chemical Name or Material 2-Amino-3,5-dibromopyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.