missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethoxythiobenzamide, 97%, Thermo Scientific™

Catalog Number 15428857
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1379207-02-9
C9H12N2O2S
212.27
MFCD16658867
MMFBQWVQXDCWDY-UHFFFAOYSA-N
2-amino-4,5-dimethoxythiobenzamide, 2-amino-4,5-dimethoxybenzothioamide
82673739
2-amino-4,5-dimethoxybenzenecarbothioamide
COC1=C(OC)C=C(C(N)=S)C(N)=C1
This item is not returnable. View return policy
1379207-02-9
C9H12N2O2S
212.27
MFCD16658867
MMFBQWVQXDCWDY-UHFFFAOYSA-N
2-amino-4,5-dimethoxythiobenzamide, 2-amino-4,5-dimethoxybenzothioamide
82673739
2-amino-4,5-dimethoxybenzenecarbothioamide
COC1=C(OC)C=C(C(N)=S)C(N)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5-dimethoxythiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'