missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)benzenethiol Hydrochloride, TRC

Product Code. 30320480
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30320480 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30320480

Brand: Toronto Research Chemicals TRCA6293305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-4-(trifluoromethyl)benzenethiol Hydrochloride
CAS 4274-38-8
Molecular Formula C7H6F3NS . HCl
Quantity 5 g
Synonym 3-Amino-4-mercaptobenzotrifluoride Hydrochloride, 4-Trifluoromethyl-2-aminothiophenol Hydrochloride, 2-Amino-α,α,α-trifluoro-p-toluenethiol Hydrochloride
InChI Formula InChI=1S/C7H6F3NS.ClH/c8-7(9,10)4-1-2-6(12)5(11)3-4;/h1-3,12H,11H2;1H
SMILES SC1=CC=C(C(F)(F)F)C=C1N.[H]Cl
IUPAC Name 2-Amino-4-(trifluoromethyl)benzenethiol Hydrochloride
Molecular Weight (g/mol) 229.65
Formula Weight 229
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.