missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-methoxypyridine, 96%, Thermo Scientific Chemicals

Product Code. 15490928
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490928 1 g 1g
15400938 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490928 Supplier Thermo Scientific Alfa Aesar Supplier No. H54723.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42409-58-5
Molecular Formula C6H7BrN2O
Molecular Weight (g/mol) 203.039
MDL Number MFCD09909849
InChI Key NFBIWMFMHLPVLT-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-methoxypyridine, 2-amino-3-methoxy-5-bromopyridine, 5-bromo-3-methoxy-pyridin-2-ylamine, 5-bromo-3-methoxy-2-pyridinamine, 5-bromo-3-methoxy-2-pyridylamine, 5-bromo-3-methoxypyridine-2-amine, 3-methoxy-5-bromopyridine-2-amine, 5-bromo-3-methoxy-pyridin-2-amine, 2-pyridinamine,5-bromo-3-methoxy, 5-bromo-3-methoxy-2-pyridyl amine
PubChem CID 22271407
IUPAC Name 5-bromo-3-methoxypyridin-2-amine
SMILES COC1=CC(=CN=C1N)Br

Specifications

Melting Point 78°C to 82°C
Quantity 1 g
Formula Weight 203.04
Percent Purity 96%
Chemical Name or Material 2-Amino-5-bromo-3-methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.