missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-fluoropyridine, 98%

Product Code. 11315075
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11315075 1 g 1g
15413877 250 mg 250mg
15403877 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11315075

Brand: Thermo Scientific Alfa Aesar H30135.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 246847-98-3 | C5H4ClFN2 | 146.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 246847-98-3
Molecular Formula C5H4ClFN2
Molecular Weight (g/mol) 146.55
MDL Number MFCD09878434
InChI Key FPUPPVDVOFJSEP-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-3-fluoropyridine, 5-chloro-3-fluoro-2-pyridinamine, 2-amino-3-fluoro-5-chloropyridine, 2-pyridinamine, 5-chloro-3-fluoro, 5-chloro-3-fluoro-2-pyridylamine, 5-chloro-3-fluoro-pyridin-2-amine, pubchem9633, acmc-1ceu9, ksc554c5t, 2-pyridinamine,5-chloro-3-fluoro
PubChem CID 11665433
IUPAC Name 5-chloro-3-fluoropyridin-2-amine
SMILES NC1=NC=C(Cl)C=C1F

Specifications

Melting Point 95°C to 97°C
Quantity 1 g
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.