missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™

Catalog Number p-7077301
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5428-89-7
C4H4ClN3
129.55
OQZMDDKDHRIGDY-UHFFFAOYSA-N
2-amino-5-chloropyrimidine, 2-pyrimidinamine, 5-chloro, 5-chloro-2-pyrimidinamine, pyrimidine, 2-amino-5-chloro, 5-chloropyrimidine-2-ylamine, 5-chlorpyrimidin-2-amin, chloropyrimidinamine, zlchem 634, pubchem9619, acmc-1aya8
79479
5-chloropyrimidin-2-amine
C1=C(C=NC(=N1)N)Cl
This item is not returnable. View return policy
5428-89-7
C4H4ClN3
129.55
OQZMDDKDHRIGDY-UHFFFAOYSA-N
2-amino-5-chloropyrimidine, 2-pyrimidinamine, 5-chloro, 5-chloro-2-pyrimidinamine, pyrimidine, 2-amino-5-chloro, 5-chloropyrimidine-2-ylamine, 5-chlorpyrimidin-2-amin, chloropyrimidinamine, zlchem 634, pubchem9619, acmc-1aya8
79479
5-chloropyrimidin-2-amine
C1=C(C=NC(=N1)N)Cl
Melting Point 235.0°C to 238.0°C
Boiling Point 314.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chloropyrimidine
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-5-chloropyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'