missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodopyrimidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7026616
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1445-39-2
C4H4IN3
221.001
MFCD01075666
HAFKCGZQRIIADX-UHFFFAOYSA-N
2-amino-5-iodopyrimidine, 2-amino-5-iodo-pyrimidine, 5-iodo-pyrimidin-2-ylamine, 5-iodopyrimidine-2-ylamine, zlchem 484, 5-iodo-2-pyrimidinamine, 2-pyrimidinamine, 5-iodo, acmc-1c050
241102
5-iodopyrimidin-2-amine
C1=C(C=NC(=N1)N)I
This item is not returnable. View return policy
1445-39-2
C4H4IN3
221.001
MFCD01075666
HAFKCGZQRIIADX-UHFFFAOYSA-N
2-amino-5-iodopyrimidine, 2-amino-5-iodo-pyrimidine, 5-iodo-pyrimidin-2-ylamine, 5-iodopyrimidine-2-ylamine, zlchem 484, 5-iodo-2-pyrimidinamine, 2-pyrimidinamine, 5-iodo, acmc-1c050
241102
5-iodopyrimidin-2-amine
C1=C(C=NC(=N1)N)I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 217°C to 220°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 221
Percent Purity 97%
Chemical Name or Material 2-Amino-5-iodopyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'