missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiobenzamide, 97%, Thermo Scientific™

Catalog Number 15408847
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
25026-97-5
C7H7N3O2S
197.212
MFCD02258912
FHYFXVGYNCGBAT-UHFFFAOYSA-N
2-amino-5-nitrothiobenzamide, benzenecarbothioamide, 2-amino-5-nitro, 2-amino-5-nitrobenzene-1-carbothioamide, 2-amino-5-nitrobenzothioamide
3034350
2-amino-5-nitrobenzenecarbothioamide
C1=CC(=C(C=C1[N+](=O)[O-])C(=S)N)N
This item is not returnable. View return policy
25026-97-5
C7H7N3O2S
197.212
MFCD02258912
FHYFXVGYNCGBAT-UHFFFAOYSA-N
2-amino-5-nitrothiobenzamide, benzenecarbothioamide, 2-amino-5-nitro, 2-amino-5-nitrobenzene-1-carbothioamide, 2-amino-5-nitrobenzothioamide
3034350
2-amino-5-nitrobenzenecarbothioamide
C1=CC(=C(C=C1[N+](=O)[O-])C(=S)N)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 197.21
Percent Purity 97%
Chemical Name or Material 2-Amino-5-nitrothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'