missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-4510252
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
96-50-4
C3H4N2S
100.14
MFCD00005325
RAIPHJJURHTUIC-UHFFFAOYSA-N
2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
2155
CHEBI:40782
1,3-thiazol-2-amine
NC1=NC=CS1
This item is not returnable. View return policy
96-50-4
C3H4N2S
100.14
MFCD00005325
RAIPHJJURHTUIC-UHFFFAOYSA-N
2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
2155
CHEBI:40782
1,3-thiazol-2-amine
NC1=NC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 86°C to 91°C
Color Brown
Boiling Point 117°C (20.0 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 100 g
Beilstein 27,155
Fieser 04,19
Merck Index 15,475
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in most common organic solvents,freely soluble in hcl and 20% h2so4
Formula Weight 100.14
Percent Purity 97%
Physical Form Crystalline Powder, Grains, Lumps or Flakes
Chemical Name or Material 2-Aminothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'