Learn More
2-(Benzimidazolylthio)acetic acid hydrazide, 97%, Thermo Scientific™
Product Code. 11433417
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433417 5 g 5g
11443417 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433417 Supplier Thermo Scientific Alfa Aesar Supplier No. A17198.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30065-27-1
Molecular Formula C9H10N4OS
Molecular Weight (g/mol) 222.27
MDL Number MFCD00171027
InChI Key UIHKXOZJFOTOCE-UHFFFAOYSA-N
Synonym 2-benzimidazolylthio acetic acid hydrazide, 2-1h-benzo d imidazol-2-yl thio acetohydrazide, 2-1h-1,3-benzodiazol-2-ylsulfanyl acetohydrazide, 2-1h-benzimidazol-2-ylthio acetohydrazide, 2-1h-benzimidazol-2-ylsulfanyl acetohydrazide, 2-1h-1,3-benzimidazol-2-ylsulfanyl acetohydrazide, 1h-benzoimidazol-2-ylsulfanyl-acetic acid hydrazide, 2-benzimidazol-2-ylthioacetohydrazide, 2-1h-benzo d imidazol-2-ylthio acetohydrazide
PubChem CID 676606
SMILES NNC(=O)CSC1=NC2=CC=CC=C2N1
Specifications
Melting Point ∼160°C (decomposition)
Quantity 5 g
IUPAC Name 2-(1H-benzimidazol-2-ylsulfanyl)acetohydrazide
Formula Weight 222.27
Percent Purity 97%
Chemical Name or Material 2-(Benzimidazolylthio)acetic acid hydrazide