missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylcyclohexanone, 97%, ACROS Organics™

Product Code. p-7077378 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15234278

Brand: Thermo Scientific Acros 451860010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 946-33-8
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
InChI Key CUYLPYBYCYQGCE-UHFFFAOYSA-N
PubChem CID 98945
IUPAC Name 2-benzylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)CC2=CC=CC=C2

Specifications

Density 1.0240g/mL
Boiling Point 103.0°C to 105.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Linear Formula C6H5CH2C6H9O
Quantity 1 g
Specific Gravity 1.024
Synonym 2-benzylcyclohexanone, cyclohexanone, 2-phenylmethyl, 2-benzyl cyclohexanone, 2-benzyl-cyclohexan-1-one, 2-phenylmethyl cyclohexanone, cyclohexanone,2-phenylmethyl, 2rs-2-phenylmethylcyclohexanone
Formula Weight 188.27
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2-Benzylcyclohexanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzylcyclohexan-1-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.