Learn More
Sartorius 2 Branch Microsart™ Manifolds
Product Code. p-7163892 Shop All Sartorius Products
Click to view available options
Type:
Biosart 100 Manifold
Microsart Manifold
Unit Size:
Each
Product Code. 16458193
Brand: Sartorius 168M2BS100
Only null left
Add to basket
Add to basket
Filtration stand designed to adapt to a variety of different needs.
  • 100% stainless steel for minimized cleaning effort
  • Full autoclavability without disassembly
  • Sterile venting in critical applications
  • Reduced risk of secondary contamination
  • Quick connection adapters for fast connection of tubing and fast interconnection of 2 manifolds
  • Low working height for ergonomic working in laminar flow
  • Accurately detects and quantifies microorganisms in liquid samples
  • Method of choice for liquid testing
  • Provides reliable and reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Manifold
For Use With (Equipment) Biosart 100 Monitor
Material Stainless Steel
Sterility Non-sterile
Quantity 1
Type Biosart 100 Manifold
Volume (Metric) 100 mL
Length (Metric) 246 mm
Width (Metric) 118 mm
Height (Metric) 130 mm