missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(difluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number 15426507
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1211515-00-2
C6H4BrF2N
208.01
MFCD18257230
RBKISNDTOFJWQZ-UHFFFAOYSA-N
2-bromo-3-difluoromethyl pyridine
56604203
2-bromo-3-(difluoromethyl)pyridine
FC(F)C1=C(Br)N=CC=C1
This item is not returnable. View return policy
1211515-00-2
C6H4BrF2N
208.01
MFCD18257230
RBKISNDTOFJWQZ-UHFFFAOYSA-N
2-bromo-3-difluoromethyl pyridine
56604203
2-bromo-3-(difluoromethyl)pyridine
FC(F)C1=C(Br)N=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-(difluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'