missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoroaniline, 97%, Thermo Scientific Chemicals

Catalog Number 15408268
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
111721-75-6
C6H5BrFN
190.015
MFCD07369915
XZRSXRUYZXBTGD-UHFFFAOYSA-N
2-bromo-3-fluorophenylamine, 2-bromo-3-fluoro-phenylamine, 2-bromo-3-fluorobenzenamine, benzenamine, 2-bromo-3-fluoro, pubchem1523, acmc-1c6kb, 2-bromo-3-fluoro-aniline, ksc493i1t, 2-bromo-3-fluoro-phenyl-amine
14770005
2-bromo-3-fluoroaniline
C1=CC(=C(C(=C1)F)Br)N
This item is not returnable. View return policy
111721-75-6
C6H5BrFN
190.015
MFCD07369915
XZRSXRUYZXBTGD-UHFFFAOYSA-N
2-bromo-3-fluorophenylamine, 2-bromo-3-fluoro-phenylamine, 2-bromo-3-fluorobenzenamine, benzenamine, 2-bromo-3-fluoro, pubchem1523, acmc-1c6kb, 2-bromo-3-fluoro-aniline, ksc493i1t, 2-bromo-3-fluoro-phenyl-amine
14770005
2-bromo-3-fluoroaniline
C1=CC(=C(C(=C1)F)Br)N

2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Density 1.670 g/mL
Flash Point 94°C (201°F)
Refractive Index 1.583
Quantity 250 mg
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-3-fluoroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'