missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methyl-5-nitropyridine, 97%

Product Code. 11369845
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11369845 1 g 1g
11379845 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11369845

Brand: Thermo Scientific Alfa Aesar L19851.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 23132-21-0 | C6H5BrN2O2 | 217.022 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23132-21-0
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD03095065
InChI Key FZIQHPKXSLHGBZ-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-3-picoline, 2-bromo-3-methyl-5-nitro-pyridine, pyridine, 2-bromo-3-methyl-5-nitro, 2-hydroxy-5-nitro-3-picoline 2-hydroxy-3-methyl-5-nitropyridine, pubchem10608, acmc-209g1c, ksc496c3r, fziqhpkxslhgbz-uhfffaoysa, 2-bromo-3-methyl-5-nitro pyridine, 2-bromanyl-3-methyl-5-nitro-pyridine
PubChem CID 5200466
IUPAC Name 2-bromo-3-methyl-5-nitropyridine
SMILES CC1=CC(=CN=C1Br)[N+](=O)[O-]

Specifications

Melting Point 55°C to 59°C
Quantity 1 g
Beilstein 1565441
Formula Weight 217.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methyl-5-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.