missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoro-4-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number 15411669
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
180608-37-1
C6H5BrFN
190.015
MFCD09839269
CIDWWTCSJWQJQR-UHFFFAOYSA-N
29919366
2-bromo-6-fluoro-4-methylpyridine
CC1=CC(=NC(=C1)Br)F
This item is not returnable. View return policy
180608-37-1
C6H5BrFN
190.015
MFCD09839269
CIDWWTCSJWQJQR-UHFFFAOYSA-N
29919366
2-bromo-6-fluoro-4-methylpyridine
CC1=CC(=NC(=C1)Br)F

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Quantity 250 mg
Beilstein 13574124
Synonym 2-bromo-6-fluoro-4-picoline, 2-bromo4-methyl6-fluoropyridine, abbypharma ap-30-7592, 2-bromo-6-fluoro-4-methyl-pyridine, pyridine, 2-bromo-6-fluoro-4-methyl, pubchem6097, 2-bromo-6-fluoro-4-methyipyridine, pyridine,2-bromo-6-fluoro-4-methyl, 2-bromo-6-fluoro-4-methylpyridine;2-bromo-4-methyl-6-fluoropyridine
Solubility Information Slightly soluble in water.
Formula Weight 190.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluoro-4-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'