missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-6'-fluoroacetophenone, 96%, Thermo Scientific Chemicals

Catalog Number 15408898
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
928715-37-1
C8H6BrFO
217.037
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluoro-phenyl ethanone, ethanone, 1-2-bromo-6-fluorophenyl
66570680
1-(2-bromo-6-fluorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Br)F
This item is not returnable. View return policy
928715-37-1
C8H6BrFO
217.037
ZTLLHQUYOLPVAR-UHFFFAOYSA-N
1-2-bromo-6-fluorophenyl ethanone, 2'-bromo-6'-fluoroacetophenone, 1-2-bromo-6-fluorophenyl ethan-1-one, 1-2-bromo-6-fluoro-phenyl ethanone, ethanone, 1-2-bromo-6-fluorophenyl
66570680
1-(2-bromo-6-fluorophenyl)ethanone
CC(=O)C1=C(C=CC=C1Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2'-Bromo-6'-fluoroacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'