missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-(3,4-dichlorobenzyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 15405746
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1421262-30-7
C13H10BrCl2NO2S
395.092
MFCD22683125
BBKPFBULBLAWNE-UHFFFAOYSA-N
2-bromo-n-3,4-dichlorophenyl methyl benzenesulfonamide, 2-bromo-n-3,4-dichlorobenzyl benzenesulfonamide
97030408
2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide
C1=CC=C(C(=C1)S(=O)(=O)NCC2=CC(=C(C=C2)Cl)Cl)Br
This item is not returnable. View return policy
1421262-30-7
C13H10BrCl2NO2S
395.092
MFCD22683125
BBKPFBULBLAWNE-UHFFFAOYSA-N
2-bromo-n-3,4-dichlorophenyl methyl benzenesulfonamide, 2-bromo-n-3,4-dichlorobenzyl benzenesulfonamide
97030408
2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide
C1=CC=C(C(=C1)S(=O)(=O)NCC2=CC(=C(C=C2)Cl)Cl)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 395.1
Percent Purity 97%
Chemical Name or Material 2-Bromo-N-(3,4-dichlorobenzyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'