missing translation for 'onlineSavingsMsg'
Learn More

2-Bromohexanoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7027458
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
616-05-7
C6H11BrO2
195.056
MFCD00004218
HZTPKMIMXLTOSK-UHFFFAOYSA-N
2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
12013
2-bromohexanoic acid
CCCCC(C(=O)O)Br
This item is not returnable. View return policy
616-05-7
C6H11BrO2
195.056
MFCD00004218
HZTPKMIMXLTOSK-UHFFFAOYSA-N
2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
12013
2-bromohexanoic acid
CCCCC(C(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3°C to 5°C
Density 1.37
Boiling Point 136°C to 138°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3265
Merck Index 14,1415
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material 2-Bromohexanoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'