missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Bromomethyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H52467.MD

Additional Details : CAS : 91983-14-1 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15409217

  • 36.87€ / 250mg


Estimated Shipment
12-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

91983-14-1
214.853
MYVJCOQGXCONPE-UHFFFAOYSA-N
2773278
B(C1=CC=CC=C1CBr)(O)O
C7H8BBrO2
MFCD01318941
2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, o-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, pubchem7781
[2-(bromomethyl)phenyl]boronic acid

Specifications

2-(Bromomethyl)benzeneboronic acid
250mg
C7H8BBrO2
2-bromomethylphenylboronic acid, 2-bromomethyl benzeneboronic acid, 2-bromomethyl phenylboronic acid, 2-bromomethyl phenyl boronic acid, 2-bromomethylphenylboronicacid, 2-boronobenzyl bromide, o-boronobenzyl bromide, 2-bromomethylphenyl boronic acid, pubchem7781
B(C1=CC=CC=C1CBr)(O)O
214.853
214.85
145°C to 148°C
91983-14-1
MFCD01318941
MYVJCOQGXCONPE-UHFFFAOYSA-N
[2-(bromomethyl)phenyl]boronic acid
2773278
97%
Videos
SDS Documents