missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopropionyl chloride, 98%

Product Code. 11450068
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11450068

Brand: Thermo Scientific Alfa Aesar B23662.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7148-74-5 | C3H4BrClO | 171.42 g/mol

2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.

Solubility
Reacts with water.

Notes
Protect from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7148-74-5
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000711
InChI Key OZGMODDEIHYPRY-UHFFFAOYNA-N
Synonym 2-bromopropionyl chloride, propanoyl chloride, 2-bromo, 2-bromopropionylchloride, 2-bromo-propionyl chloride, 2-brominepropionylchloride, alpha-bromopropionyl chloride, 2-bromopropionic acid chloride, sgqdjlpqbsmuh@, 2-bromopropanoylchloride, acmc-1cjhb
PubChem CID 97980
IUPAC Name 2-bromopropanoyl chloride
SMILES CC(Br)C(Cl)=O

Specifications

Density 1.7
Boiling Point 131°C to 133°C
Flash Point 51°C (125°F)
Refractive Index 1.478
Quantity 10 g
UN Number UN2920
Beilstein 773800
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 171.43
Percent Purity 98%
Chemical Name or Material 2-Bromopropionyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.