missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™

Catalog Number p-7120057
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -87°C
Density 0.805g/mL at 25°C
Boiling Point 80°C
Quantity 500 mL
Flash Point −3°C (26.6°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index n20/D 1.379
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'