missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Riedel-de Haen™ 2-Butanone, puriss. p.a. ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haen™

Puriss. p.a., ACS reagent,≥99.5% (GC)

Brand:  Honeywell Riedel-de Haen™ 04380-250ML

Additional Details : CAS : 78-93-3 Poids : 0.20100kg Transport : UN number : 1193 Chem class : 3 Pack group : II

Packaging Glass Bottle
Quantity 250mL

 View more versions of this product

Product Code. 15650760

  • 17.15€ / 250mL


Expédition estimée
17-02-2021
Add to basket

Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

Specifications

Colorless
Glass Bottle
−3°C (26.6°F)
-87°C
0.805g/mL at 25°C
n20/D 1.379(lit.)
79°C to 81°C
250mL
Videos
SDS Documents