missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)benzyl alcohol, 97+%, Thermo Scientific™

Product Code. 11440817
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11440817 5 g 5g
11430817 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11440817

Brand: Thermo Scientific Alfa Aesar B23212.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261763-20-6
Molecular Formula C8H6ClF3O
Molecular Weight (g/mol) 210.58
MDL Number MFCD01631537
InChI Key MQUXXVLMXCHGDZ-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl benzyl alcohol, 2-chloro-3-trifluoromethyl phenyl methanol, 2-chloro-3-trifluoromethyl benzylalcohol, 2-chloro-3-trifluoromethyl phenyl methan-1-ol, acmc-1coey, rarechem al bd 0475, rarechem al bd 1301, 2-chloro-3-trifluoromethyl-benzylalcohol, 2-chloro-3-trifluoromethylbenzyl alcohol
PubChem CID 2778108
IUPAC Name [2-chloro-3-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=C(C(=C1)C(F)(F)F)Cl)CO

Specifications

Melting Point 74°C to 77°C
Quantity 5 g
Formula Weight 210.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.