Learn More
2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine, 97%, Alfa Aesar™
Brand: Alfa Aesar H63212.03
Additional Details : CAS : 372183-70-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
372183-70-5 | |
250.701 | |
VALSYGYSZKIHMQ-UHFFFAOYSA-N | |
53411892 | |
CC1=CC(=NC(=N1)Cl)CCC2=CC=C(C=C2)F |
C13H12ClFN2 | |
MFCD04974533 | |
2-chloro-4-2-4-fluorophenyl ethyl-6-methylpyrimidine, 2-chloro-4-2-4-fluorophenyl-ethyl-6-methylpyrimidine, c13h13clfn3, 2-chloro-4-4-fluorophenethyl-6-methylpyrimidine | |
2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine |
Specifications
2-Chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine | |
372183-70-5 | |
MFCD04974533 | |
VALSYGYSZKIHMQ-UHFFFAOYSA-N | |
2-chloro-4-[2-(4-fluorophenyl)ethyl]-6-methylpyrimidine | |
53411892 | |
Solid |
1g | |
C13H12ClFN2 | |
2-chloro-4-2-4-fluorophenyl ethyl-6-methylpyrimidine, 2-chloro-4-2-4-fluorophenyl-ethyl-6-methylpyrimidine, c13h13clfn3, 2-chloro-4-4-fluorophenethyl-6-methylpyrimidine | |
CC1=CC(=NC(=N1)Cl)CCC2=CC=C(C=C2)F | |
250.701 | |
250.07 | |
97% |