missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
Solubility
Soluble in acetonitrile, dichloromethane, tetrahydrofuron and dimethylformamide.
Notes
Light and heat sensitive. Incompatible with water and strong oxidizing agents.
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
Solubility
Soluble in acetonitrile, dichloromethane, tetrahydrofuron and dimethylformamide.
Notes
Light and heat sensitive. Incompatible with water and strong oxidizing agents.
Specifications
Melting Point | 36°C to 40°C |
Boiling Point | 127°C to 128°C (11 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in acetonitrile,dichloromethane,tetrahydrofuron and dimethylformamide. |
Formula Weight | 202.54 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'