missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15411186
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1449132-57-3
C13H9BCl3NO3
344.379
MFCD22125068
LGHQYAUSSYHBTK-UHFFFAOYSA-N
2-chloro-5-2,5-dichlorophenylcarbamoyl benzeneboronic acid, 2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid, 2-chloro-5-2,5-dichlorophenylcarbamoyl phenylboronic acid
73995808
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
This item is not returnable. View return policy
1449132-57-3
C13H9BCl3NO3
344.379
MFCD22125068
LGHQYAUSSYHBTK-UHFFFAOYSA-N
2-chloro-5-2,5-dichlorophenylcarbamoyl benzeneboronic acid, 2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid, 2-chloro-5-2,5-dichlorophenylcarbamoyl phenylboronic acid
73995808
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 344.39
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'