missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(3-nitrophenylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7037265
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096341-27-2
C13H10BClN2O5
320.49
MFCD22125186
LEIFVWJAWHHHHN-UHFFFAOYSA-N
2-chloro-5-3-nitrophenylcarbamoyl phenylboronic acid, 2-chloro-5-3-nitrophenylcarbamoyl benzeneboronic acid
73996201
This item is not returnable. View return policy
2096341-27-2
C13H10BClN2O5
320.49
MFCD22125186
LEIFVWJAWHHHHN-UHFFFAOYSA-N
2-chloro-5-3-nitrophenylcarbamoyl phenylboronic acid, 2-chloro-5-3-nitrophenylcarbamoyl benzeneboronic acid
73996201

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 320.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(3-nitrophenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'