missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15412346
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096332-75-9
C13H9BClFN2O5
338.48
MFCD20040332
HBBVKWQZDSTEMK-UHFFFAOYSA-N
2-chloro-5-4-fluoro-2-nitrophenylcarbamoyl phenylboronic acid, 2-chloro-5-4-fluoro-2-nitrophenylcarbamoyl benzeneboronic acid
73996193
This item is not returnable. View return policy
2096332-75-9
C13H9BClFN2O5
338.48
MFCD20040332
HBBVKWQZDSTEMK-UHFFFAOYSA-N
2-chloro-5-4-fluoro-2-nitrophenylcarbamoyl phenylboronic acid, 2-chloro-5-4-fluoro-2-nitrophenylcarbamoyl benzeneboronic acid
73996193

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 338.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'