missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%

Brand:  Alfa Aesar™ H51035.06

Additional Details : CAS : 7298-89-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407927

  • 847.00€ / 5g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Applications
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

7298-89-7
174.624
VOBIHUAWDXUCPH-UHFFFAOYSA-N
122278
CC1(CC(=O)C(C(=O)C1)Cl)C
C8H11ClO2
MFCD00010502
chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon
2-chloro-5,5-dimethylcyclohexane-1,3-dione

Specifications

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
161°C to 164°C
7298-89-7
MFCD00010502
chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon
VOBIHUAWDXUCPH-UHFFFAOYSA-N
2-chloro-5,5-dimethylcyclohexane-1,3-dione
122278
98%
Moisture sensitive
5g
C8H11ClO2
2326749
Insoluble in water.
CC1(CC(=O)C(C(=O)C1)Cl)C
174.624
174.62
Videos
SDS Documents