Learn More
2-Chloro-6-fluoro-3-methoxyaniline, 97%, Thermo Scientific™
Product Code. 15485497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15485497
Brand: Thermo Scientific Alfa Aesar H32742.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017777-58-0
Molecular Formula C7H7ClFNO
Molecular Weight (g/mol) 175.587
MDL Number MFCD09832265
InChI Key QMCMJQAVMDONLJ-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-m-anisidine, 2-chloro-6-fluoro-3-methoxy-aniline, 2-chloro-6-fluoro-3-methoxybenzenamine, 2-chloro-6-fluoro-3-methoxyphenylamine
PubChem CID 18444747
IUPAC Name 2-chloro-6-fluoro-3-methoxyaniline
SMILES COC1=C(C(=C(C=C1)F)N)Cl
Specifications
Melting Point 37°C to 40°C
Quantity 1 g
UN Number UN2811
Formula Weight 175.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluoro-3-methoxyaniline