missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-N,N-dimethylacetamide, 97%, Thermo Scientific Chemicals

Catalog Number p-4228504
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2675-89-0
C4H8ClNO
121.57
XBPPLECAZBTMMK-UHFFFAOYSA-N
n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
75886
2-chloro-N,N-dimethylacetamide
CN(C)C(=O)CCl
This item is not returnable. View return policy
2675-89-0
C4H8ClNO
121.57
XBPPLECAZBTMMK-UHFFFAOYSA-N
n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
75886
2-chloro-N,N-dimethylacetamide
CN(C)C(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Colorless to Yellow
Density 1.1820g/mL
Boiling Point 98°C to 100°C (11.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CON(CH3)2
Quantity 5 mL
Specific Gravity 1.182
Solubility Information Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble
Formula Weight 121.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-N, N-dimethylacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-N,N-dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'